Fisherbrand™ Tissue Path™ IV Tissue Cassettes
Contain no jagged edges or uneven surfaces
$583.31 - $1953.22
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
22-272416
|
Fisherbrand™
58409 |
White |
Case of 1000 Each for $596.22
|
|
|||||
22-272420
|
Fisherbrand™
58413 |
Pink |
Case of 1000 Each for $583.31
|
|
|||||
22-272419
|
Fisherbrand™
58412 |
Green |
Case of 1000 Each for $588.59
|
|
|||||
22-272417
|
Fisherbrand™
58410 |
Blue |
Case of 1000 Each for $596.22
|
|
|||||
22-272418
|
Fisherbrand™
58411 |
Yellow |
Case of 1000 Each for $590.78
|
|
|||||
22-272421
|
Fisherbrand™
58414 |
Tan |
Case of 1000 Each for $645.28
|
|
|||||
22-272422
|
Fisherbrand™
58415 |
Peach |
Case of 1000 Each for $1,001.09
|
|
|||||
22-272423
|
Fisherbrand™
58416 |
Gray |
Case of 1000 Each for $1,018.94
|
|
|||||
22-292443
|
Fisherbrand™
58164 |
Gold |
Case of 1000 Each for $1,953.22
|
|
|||||
22-292444
|
Fisherbrand™
58165 |
Lilac |
Case of 1000 Each for $585.25
|
|
|||||
Description
- Unique, two-piece molded hinge ensures specimen safety and facilitates easy removal of the cassette cover from the base during the embedding procedure
- Cover and base can be disassembled and reassembled with ease
- Cassettes fit all sizes of popular base molds, metal and plastic
- Three-sided writing surface assures positive identification
- Slotted design ensures thorough and rapid exchange of processing fluids
- Resistant to solvents, decalcifying solutions, and compatible with microwave techniques
Order Info
- Supplied in cases of four handy dispenser boxes, 250 per box
Specifications
Slots | |
Plastic | |
Tissue Cassette |