missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ Capitol Vial Transport Tubes

Caps and tubes remain leak-resistant even when vacuum-packed or pressurized.

Supplier:  Thermo Scientific™ S40100

 View more versions of this product

Catalog No. 22-038966


Explore available promotions

Description

Description

  • 10mL Polypropylene tubes
  • Rounded bottoms with freestanding skirts
  • Tubes are etched with graduations from 1.0 to 8.0mL, in 1.0mL increments

Order Info

Clinical trials serum transport tubes come precapped; caps for specimen transport tubes are sold separately; packaged in cases of 2000

Specifications

Specifications

Clear
Polypropylene
10 mL
Round
Not Included
2000 Cs.
15 mm
Non-sterile
1 to 8 mL
0.33 oz.
86 mm
DOT, IATA
Videos
SDS
Documents

Documents

Promotions

Promotions