missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Thermo Scientific™ Capitol Vial Transport Tubes
Caps and tubes remain leak-resistant even when vacuum-packed or pressurized.
$503.81 - $1070.72
Specifications
Color | Clear |
---|---|
Diameter (Metric) Outer | 15 mm |
Material | Polypropylene |
Sterility | Non-sterile |
Volume (Metric) | 10 mL |
Description
- 10mL Polypropylene tubes
- Rounded bottoms with freestanding skirts
- Tubes are etched with graduations from 1.0 to 8.0mL, in 1.0mL increments
Order Info
Clinical trials serum transport tubes come precapped; caps for specimen transport tubes are sold separately; packaged in cases of 2000
Specifications
Clear | |
Polypropylene | |
10 mL | |
Round | |
86 mm |
15 mm | |
Non-sterile | |
1 to 8 mL | |
0.33 oz. | |
2000 Cs. |