Fisherbrand™ UriSystem™ Caps, Blue
Choose from five colors
 View more versions of this product
Catalog No. 14-377-255
Explore available promotions
Description
Description
Choose from five colors
  • Tight-fitting plastic caps for standard UriSystem tubes (14-377-252 and 14-377-253)
  • Prevent spilling and contamination
Specifications
Specifications
UriSystem Cap
Neon Orange
For standard UriSystem tubes
Plastic
Prevents spilling and contamination