Description
- Firm, positive seal by sealing either inside the tube or over the tube rim
- One-size design seals most 16, 13, 12 and 10mm blood collection, glass and plastic test tubes
- No need to purchase size-specific closures
- Help with laboratory standardization and managed cost initiatives
- One-hand technique, seal under various temperature/storage conditions, help preserve sample integrity and minimize evaporation loss
Specifications
Not Autoclavable | |
Blue | |
16, 13, 12 and 10 mm Blood Collection, Glass and Plastic Test Tube |
LDPE | |
Non-sterile |