Fisherbrand™ VersaClosure™ Tube Closures
Universal cap closure
$117.43 - $176.46
Specifications
Autoclavable | Not Autoclavable |
---|---|
Material | LDPE |
Sterility | Non-sterile |
For Use With (Equipment) | 16, 13, 12 and 10 mm Blood Collection, Glass and Plastic Test Tube |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
02-707-22
|
Fisherbrand™
A20010FIS |
Gray |
Pack of 1000 Each for $117.43
Case of 10 Pack for $1,174.30
|
|
|||||
02-707-17
|
N/A | Clear |
Pack of 1000 Each for $170.19
Case of 10 Pack for $1,701.90
|
|
|||||
02-707-18
|
N/A | Red |
Pack of 1000 Each for $176.46
Case of 10 Pack for $1,764.60
|
|
|||||
02-707-19
|
N/A | Lavender |
Pack of 1000 Each for $124.81
Case of 10 Pack for $1,248.10
|
|
|||||
02-707-20
|
N/A | Green |
Pack of 1000 Each for $176.46
Case of 10 Pack for $1,764.60
|
|
|||||
02-707-21
|
N/A | Blue |
Pack of 1000 Each for $171.23
Case of 10 Pack for $1,712.30
|
|
|||||
02-707-23
|
N/A | Yellow |
Pack of 1000 Each for $171.23
Case of 10 Pack for $1,712.30
|
|
|||||
02-707-24
|
N/A | Black |
Pack of 1000 Each for $171.23
Case of 10 Pack for $1,712.30
|
|
|||||
Description
- Firm, positive seal by sealing either inside the tube or over the tube rim
- One-size design seals most 16, 13, 12 and 10mm blood collection, glass and plastic test tubes
- No need to purchase size-specific closures
- Help with laboratory standardization and managed cost initiatives
- One-hand technique, seal under various temperature/storage conditions, help preserve sample integrity and minimize evaporation loss
Specifications
Not Autoclavable | |
LDPE | |
Non-sterile | |
16, 13, 12 and 10 mm Blood Collection, Glass and Plastic Test Tube |