Fisherbrand™ VersaClosure™ Tube Closures
Universal cap closure
 View more versions of this product
Catalog No. 02-707-23
Explore available promotions
Description
Description
  • Firm, positive seal by sealing either inside the tube or over the tube rim
  • One-size design seals most 16, 13, 12 and 10mm blood collection, glass and plastic test tubes
  • No need to purchase size-specific closures
  • Help with laboratory standardization and managed cost initiatives
  • One-hand technique, seal under various temperature/storage conditions, help preserve sample integrity and minimize evaporation loss
Specifications
Specifications
Not Autoclavable
Yellow
16, 13, 12 and 10 mm Blood Collection, Glass and Plastic Test Tube
LDPE
Non-sterile