Fisherbrand™ Premium Frosted Microscope Slides, Superfrost marking area Edge
Edge
With Superfrost™ marking area on one end, one side
 View more versions of this product
Catalog No. 12-544-2
Edge
Explore available promotions
Description
Description
With Superfrost™ marking area on one end, one side
  • Excellent retention of markings in all normal procedures
Specifications
Specifications
Microscope Slide
75 mm
3 in.
75 x 25 mm
SuperFrost Marking Area 1 End, 1 Side
25 mm
1 in.