Fischer Technical Company™ Flip Style Two Valve Natural Rubber Safety Pipet Filler
Safety Pipet Filler is easy to use and clean.
Supplier:  Fischer Technical Company™ S32247
 View more versions of this product
Catalog No. S32247
Explore available promotions
Alert:
Prop 65: Cancer and Reproductive Harm.
Description
Description
This patented ball fits pipets up to 50 mL.
  • Press on the bulb to evacuate
  • Press on the up arrow to intake solution
  • Drain by pressing on the down arrow
  • Top valve can be removed and reinserted for easy cleaning of the inside
Specifications
Specifications
Not Autoclavable
Reusable
Round
Flip
Non-sterile
50 mL
Red
Pipette
Natural Rubber
1
Pipette Filler