missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluorene, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 156131000

 View more versions of this product

Catalog No. AC156131000


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Fluorene
86-73-7
Brown to White
298.0°C
Authentic
Plastic bottle
MFCD00001111
05, 625
15, 4187
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene and carbon disu,freely soluble in glacial acetic acid
C1C2=CC=CC=C2C2=CC=CC=C12
166.22
CHEBI:28266
98+%
98%
112.0°C to 116.0°C
1.2000g/mL
151°C
98% min. (GC)
C13H10
100 g
1.2
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
NIHNNTQXNPWCJQ-UHFFFAOYSA-N
9H-fluorene
6853
166.22
Crystalline Powder

Chemical Identifiers

86-73-7
166.22
NIHNNTQXNPWCJQ-UHFFFAOYSA-N
6853
9H-fluorene
C13H10
MFCD00001111
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
CHEBI:28266
C1C2=CC=CC=C2C2=CC=CC=C12
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

EINECSNumber : 201-695-5

RTECSNumber : LL5670000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only