missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Fluorene, 98+%, Thermo Scientific Chemicals

$73.17 - $524.73

Chemical Identifiers

CAS 86-73-7
Molecular Formula C13H10
Molecular Weight (g/mol) 166.22
MDL Number MFCD00001111
InChI Key NIHNNTQXNPWCJQ-UHFFFAOYSA-N
Synonym fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
PubChem CID 6853
ChEBI CHEBI:28266
IUPAC Name 9H-fluorene
SMILES C1C2=CC=CC=C2C2=CC=CC=C12
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC156130250
SDS View Documents
Thermo Scientific Chemicals
156130250
25 g Plastic bottle
Each for $73.17
 
AC156131000
SDS View Documents
Thermo Scientific Chemicals
156131000
100 g Plastic bottle
Each for $157.86
 
AC156135000
SDS View Documents
Thermo Scientific Chemicals
156135000
500 g Plastic bottle
Each for $524.73
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

86-73-7
166.22
NIHNNTQXNPWCJQ-UHFFFAOYSA-N
6853
9H-fluorene
C13H10
MFCD00001111
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
CHEBI:28266
C1C2=CC=CC=C2C2=CC=CC=C12

Specifications

86-73-7
Brown to White
298.0°C
Authentic
Plastic bottle
MFCD00001111
05, 625
15, 4187
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene and carbon disu,freely soluble in glacial acetic acid
C1C2=CC=CC=C2C2=CC=CC=C12
166.22
CHEBI:28266
98+%
Fluorene, 98%
112.0°C to 116.0°C
1.2000g/mL
151°C
98% min. (GC)
C13H10
25 g
1.2
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene
NIHNNTQXNPWCJQ-UHFFFAOYSA-N
9H-fluorene
6853
166.22
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

EINECSNumber : 201-695-5

RTECSNumber : LL5670000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only