missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Fluorene, 98+%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 156135000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Fluorene | |
86-73-7 | |
Brown to White | |
298.0°C | |
Authentic | |
Plastic bottle | |
MFCD00001111 | |
05, 625 | |
15, 4187 | |
Solubility in water: insoluble. Other solubilities: soluble in alcohol,ether,benzene and carbon disu,freely soluble in glacial acetic acid | |
C1C2=CC=CC=C2C2=CC=CC=C12 | |
166.22 | |
CHEBI:28266 | |
98+% |
98% | |
112.0°C to 116.0°C | |
1.2000g/mL | |
151°C | |
98% min. (GC) | |
C13H10 | |
500 g | |
1.2 | |
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene | |
NIHNNTQXNPWCJQ-UHFFFAOYSA-N | |
9H-fluorene | |
6853 | |
166.22 | |
Crystalline Powder |
Chemical Identifiers
86-73-7 | |
166.22 | |
NIHNNTQXNPWCJQ-UHFFFAOYSA-N | |
6853 | |
9H-fluorene |
C13H10 | |
MFCD00001111 | |
fluorene, diphenylenemethane, o-biphenylenemethane, 2,3-benzindene, 2,2'-methylenebiphenyl, o-biphenylmethane, methane, diphenylene, fluoren, fluorenyl radical, flourene | |
CHEBI:28266 | |
C1C2=CC=CC=C2C2=CC=CC=C12 |
Safety and Handling
GHS H Statement
Very toxic to aquatic life with long lasting effects.
GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.
GHS Signal Word: Warning
EINECSNumber : 201-695-5
RTECSNumber : LL5670000
TSCA : TSCA
RUO – Research Use Only