missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One FLUOTRAC™ 200 96-well Half Area Microplate

Feature micro-thin μClear™ bottoms for exceptional optical properties.

Supplier:  Greiner Bio-One 675096

 View more versions of this product

Catalog No. 07000093


Explore available promotions

Description

Description

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Can be traced back to production through defined LOT number system
  • Reduction of sample volume by up to 50 %
  • Sta
Specifications

Specifications

Free of detectable DNase/RNase, human DNA and pyrogens.
8 x 12
Medium Binding
Polystyrene
15 to 17 μL
Medium Binding
10
128 x 85 x 14 mm
Half-Area
Not Autoclavable
Black
96
Non-sterile
40
40/Cs.
Videos
SDS
Documents

Documents

Promotions

Promotions