missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents

Supplier:  Fisher BioReagents BPBP53125

 View more versions of this product

Catalog No. BP531-25



Explore available promotions

Description

Description

Molecular Biology-grade Formaldehyde is used for denaturing RNA.
Specifications

Specifications

Formaldehyde
50-00-0,67-56-1,7732-18-5
Colorless
3 to 4.2
0.005% max.
Glass Bottle
HCHO
25 mL
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
C=O
30.03
CHEBI:16842
2mbar at 20┬░C
Molecular Biology
37% by Weight
−15°C
1.083g/cm┬│
97°C
36.5 to 38.0 %
CH2O
MFCD00003274
15, 4263
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
30.02
36.5 to 38.0%
Liquid

Chemical Identifiers

50-00-0
30.03
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00003274
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
CHEBI:16842
C=O
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

Recommended Storage : RT

SDS Documents

Documents

Promotions

Promotions