Learn More
Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents
$67.25 - $75.08
Chemical Identifiers
CAS | 50-00-0 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.03 |
MDL Number | MFCD00003274 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
PubChem CID | 712 |
ChEBI | CHEBI:16842 |
IUPAC Name | formaldehyde |
SMILES | C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
BP531-25
|
Fisher BioReagents
BPBP53125 |
25 mL | Glass Bottle |
Each for $67.25
Case of 6 Each for $403.50
|
|
||||
BP531-500
|
Fisher BioReagents
BPBP531500 |
500 mL | HDPE Bottle |
Each for $75.08
Case of 6 Each for $450.48
|
|
||||
Description
Molecular Biology-grade Formaldehyde is used for denaturing RNA.Chemical Identifiers
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Specifications
50-00-0,67-56-1,7732-18-5 | |
Colorless | |
3 to 4.2 | |
0.005% max. | |
Glass Bottle | |
HCHO | |
25 mL | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.03 | |
CHEBI:16842 | |
2mbar at 20â¬âC | |
Molecular Biology | |
Formaldehyde, 37% by Weight |
−15°C | |
1.083g/cmâ¬â | |
97°C | |
36.5 to 38.0 % | |
CH2O | |
MFCD00003274 | |
15, 4263 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
30.02 | |
36.5 to 38.0% | |
Liquid |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.
NFPA
Health:3
Flammability:2
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid
Recommended Storage : RT