Learn More
Formaldehyde (37% by Weight/Molecular Biology), Fisher BioReagents
Supplier: Fisher BioReagents BPBP531500
Description
Molecular Biology-grade Formaldehyde is used for denaturing RNA.Specifications
Formaldehyde | |
50-00-0,67-56-1,7732-18-5 | |
Colorless | |
3 to 4.2 | |
0.005% max. | |
HDPE Bottle | |
HCHO | |
500 mL | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.03 | |
CHEBI:16842 | |
2mbar at 20â¬âC | |
Molecular Biology |
37% by Weight | |
−15°C | |
1.083g/cmâ¬â | |
97°C | |
36.5 to 38.0 % | |
CH2O | |
MFCD00003274 | |
15, 4263 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
30.02 | |
36.5 to 38.0% | |
Liquid |
Chemical Identifiers
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.
NFPA
Health:3
Flammability:2
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid
Recommended Storage : RT