missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formaldehyde, 37% by Weight (With Preservative/Certified ACS), Fisher Chemical™

Stabilized with 10-15% methanol

Supplier:  Fisher Chemical F79200

 View more versions of this product

Catalog No. F79-200


Explore available promotions

Description

Description

Specifications

Specifications

Formaldehyde, 37% by Weight
50-00-0,67-56-1,7732-18-5
Colorless
0.005% max.
Poly Drum
HCHO
200 L
WSFSSNUMVMOOMR-UHFFFAOYSA-N
formaldehyde
712
30.03
Certified ACS
Liquid
With Preservative
0°C
101°C
36.5 to 38.0 %
CH2O
MFCD00003274
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
C=O
30.03
CHEBI:16842
36.5 to 38.0%
Stabilized with 10-15% Methanol

Chemical Identifiers

50-00-0
30.03
WSFSSNUMVMOOMR-UHFFFAOYSA-N
712
formaldehyde
CH2O
MFCD00003274
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde
CHEBI:16842
C=O
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.

NFPA

Health:3
Flammability:2
Instability:0

DOTInformation : DOT Class 3, : Flammable Liquid

SDS Documents

Documents

Accessories
Promotions

Promotions