Learn More
Formaldehyde, 37% by Weight (With Preservative/Certified ACS), Fisher Chemical™
Stabilized with 10-15% methanol
$298.27 - $9864.91
Identifiants chimiques
CAS | 50-00-0 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.03 |
MDL Number | MFCD00003274 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
PubChem CID | 712 |
ChEBI | CHEBI:16842 |
IUPAC Name | formaldehyde |
SMILES | C=O |
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
---|---|---|---|---|---|---|---|---|---|
Numéro de catalogue | Numéro du manufacturier. | Quantity | Packaging | Prix | Quantité | ||||
F79-1
|
Thermo Fisher Scientific
F791 |
1 L | HDPE Bottle |
chaque for $298.27
caisse de 6 chaque for $1,789.62
|
|
||||
F79P-4
|
Thermo Fisher Scientific
F79P4 |
4 L | Poly Bottle |
chaque for $363.18
caisse de 4 chaque for $1,452.72
|
|
||||
F79-200
|
Thermo Fisher Scientific
F79200 |
200 L | Poly Drum |
chaque for $9,864.91
|
|
||||
Identifiants chimiques
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Spécifications
50-00-0,67-56-1,7732-18-5 | |
Colorless | |
0.005% max. | |
HDPE Bottle | |
HCHO | |
1 L | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
30.03 | |
Certified ACS | |
Liquid |
0°C | |
101°C | |
36.5 to 38.0 % | |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.03 | |
CHEBI:16842 | |
36.5 to 38.0% | |
Stabilized with 10-15% Methanol | |
Formaldehyde, 37% by Weight, With Preservative |
Sécurité et manipulation
DANGER!
Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.
NFPA
Health:3
Flammability:2
Instability:0
missing translation for 'dotInformation' : DOT Class 3, : Flammable Liquid