missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 150690050

 View more versions of this product

Catalog No. AC150690050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Formaldehyde sodium bisulfite addition compound
870-72-4
Authentic
Glass Bottle
HOCH2SO3Na
5 g
UOULCEYHQNCFFH-UHFFFAOYSA-M
sodium;hydroxymethanesulfonate
2723822
≥94%
95%
White
95%
CH3NaO4S
MFCD00044664
sodium hydroxymethanesulfonate, formaldehyde sodium bisulfite, formbis, sodium formaldehyde bisulfite, sodium hydroxymethanesulphonate, methanesulfonic acid, hydroxy-, monosodium salt, sodium hydroxymethane sulfonate, sodium formaldehydebisulfite, unii-i566kha05f, methylolsulfonic acid sodium salt
C(O)S(=O)(=O)[O-].[Na+]
134.081
134.08
Powder

Chemical Identifiers

870-72-4
134.081
UOULCEYHQNCFFH-UHFFFAOYSA-M
2723822
C(O)S(=O)(=O)[O-].[Na+]
CH3NaO4S
MFCD00044664
sodium hydroxymethanesulfonate, formaldehyde sodium bisulfite, formbis, sodium formaldehyde bisulfite, sodium hydroxymethanesulphonate, methanesulfonic acid, hydroxy-, monosodium salt, sodium hydroxymethane sulfonate, sodium formaldehydebisulfite, unii-i566kha05f, methylolsulfonic acid sodium salt
sodium;hydroxymethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 212-800-9

RTECSNumber : PB2200000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only