missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formaldehyde Sodium Bisulfite Addition Compound 95%, Thermo Scientific Chemicals

$38.64 - $38.64

Chemical Identifiers

CAS 870-72-4
Molecular Formula CH3NaO4S
Molecular Weight (g/mol) 134.081
MDL Number MFCD00044664
InChI Key UOULCEYHQNCFFH-UHFFFAOYSA-M
Synonym sodium hydroxymethanesulfonate, formaldehyde sodium bisulfite, formbis, sodium formaldehyde bisulfite, sodium hydroxymethanesulphonate, methanesulfonic acid, hydroxy-, monosodium salt, sodium hydroxymethane sulfonate, sodium formaldehydebisulfite, unii-i566kha05f, methylolsulfonic acid sodium salt
PubChem CID 2723822
IUPAC Name sodium;hydroxymethanesulfonate
SMILES C(O)S(=O)(=O)[O-].[Na+]
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC150690050
SDS View Documents
Thermo Scientific Chemicals
150690050
5 g Glass Bottle N/A
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

870-72-4
134.081
UOULCEYHQNCFFH-UHFFFAOYSA-M
2723822
C(O)S(=O)(=O)[O-].[Na+]
CH3NaO4S
MFCD00044664
sodium hydroxymethanesulfonate, formaldehyde sodium bisulfite, formbis, sodium formaldehyde bisulfite, sodium hydroxymethanesulphonate, methanesulfonic acid, hydroxy-, monosodium salt, sodium hydroxymethane sulfonate, sodium formaldehydebisulfite, unii-i566kha05f, methylolsulfonic acid sodium salt
sodium;hydroxymethanesulfonate

Specifications

870-72-4
Authentic
Glass Bottle
HOCH2SO3Na
5 g
UOULCEYHQNCFFH-UHFFFAOYSA-M
sodium;hydroxymethanesulfonate
2723822
≥94%
Formaldehyde sodium bisulfite addition compound, 95%
White
95%
CH3NaO4S
MFCD00044664
sodium hydroxymethanesulfonate, formaldehyde sodium bisulfite, formbis, sodium formaldehyde bisulfite, sodium hydroxymethanesulphonate, methanesulfonic acid, hydroxy-, monosodium salt, sodium hydroxymethane sulfonate, sodium formaldehydebisulfite, unii-i566kha05f, methylolsulfonic acid sodium salt
C(O)S(=O)(=O)[O-].[Na+]
134.081
134.08
Powder
Videos
Safety and Handling

Safety and Handling

HYGROSCOPIC

EINECSNumber : 212-800-9

RTECSNumber : PB2200000

TSCA : TSCA

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only