Learn More

Formaldehyde (USP), Fisher Chemical™
Supplier: Fisher Chemical F77P4
Specifications
Formaldehyde, 40% by Volume | |
Pass Test | |
Colorless | |
36.5 to 38.0 % | |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.03 | |
CHEBI:16842 | |
≥34.5% | |
40% by Volume |
50-00-0,67-56-1,7732-18-5 | |
0°C | |
101°C | |
Poly Bottle | |
HCHO | |
4 L | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
30.03 | |
USP | |
Liquid |
Chemical Identifiers
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Safety and Handling

DANGER!
Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.
NFPA
Health:3
Flammability:2
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid