Learn More
Formaldehyde (USP), Fisher Chemical™
$355.69 - $11390.31
Chemical Identifiers
CAS | 50-00-0 |
---|---|
Molecular Formula | CH2O |
Molecular Weight (g/mol) | 30.03 |
MDL Number | MFCD00003274 |
InChI Key | WSFSSNUMVMOOMR-UHFFFAOYSA-N |
Synonym | formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde |
PubChem CID | 712 |
ChEBI | CHEBI:16842 |
IUPAC Name | formaldehyde |
SMILES | C=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
F77P-4
|
Thermo Fisher Scientific
F77P4 |
4 L | Poly Bottle |
Each for $355.69
Case of 4 Each for $1,422.76
|
|
||||
F77-20
|
Thermo Fisher Scientific
F7720 |
20 L | Poly Pail |
Each for $1,348.77
|
|
||||
F77P-20
|
Thermo Fisher Scientific
F77P20 |
20 L | PolyPac™ |
Each for $1,256.31
|
|
||||
F77-200
|
Thermo Fisher Scientific
F77200 |
200 L | Steel Drum |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
50-00-0 | |
30.03 | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
712 | |
formaldehyde |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
CHEBI:16842 | |
C=O |
Specifications
50-00-0,67-56-1,7732-18-5 | |
0°C | |
101°C | |
Poly Bottle | |
HCHO | |
4 L | |
WSFSSNUMVMOOMR-UHFFFAOYSA-N | |
formaldehyde | |
712 | |
30.03 | |
USP | |
Liquid |
Pass Test | |
Colorless | |
36.5 to 38.0 % | |
CH2O | |
MFCD00003274 | |
formalin, methanal, formol, methylene oxide, paraformaldehyde, oxomethane, paraform, formic aldehyde, oxymethylene, methyl aldehyde | |
C=O | |
30.03 | |
CHEBI:16842 | |
≥34.5% | |
40% by Volume | |
Formaldehyde, 40% by Volume |
Safety and Handling
DANGER!
Emergency Overview
Flammable liquid and vapor. Poison, may be fatal or cause blindness if swallowed. Cannot be made non-poisonous. Toxic by inhalation, in contact with skin and if swallowed. Causes burns by all exposure routes. Vapor harmful. May cause an allergic skin reaction. May cause central nervous system effects. WARNING! This product contains a chemical known in the State of California to cause birth defects or. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes.
NFPA
Health:3
Flammability:2
Instability:0
DOTInformation : DOT Class 3, : Flammable Liquid