missing translation for 'onlineSavingsMsg'
Learn More

Formic acid, 98+%, pure

CAS: 64-18-6 | CH2O2 | 46.025 g/mol

Supplier:  Thermo Scientific Chemicals 147932500

 Afficher plus de versions de ce produit

Numéro de catalogue. AC147932500


Il en reste null
Découvrez plus d'offres spéciales

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Spécifications

Spécifications

Formic acid
98+%
HCO2H
01,404; 02,202; 03,147; 04,239; 05,316; 07,160; 08,232; 09,226; 11,243; 13,107; 16,177
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
C(=O)O
46.025
CHEBI:30751
≥98.0%
1.2200g/mL
Authentic
Liquid
44mbar at 20°C
64-18-6
CH2O2
02, 8
15, 4269
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
284
46.02
Pure
250 mL
Plastic bottle
1.3701 to 1.3721 (20°C, 589nm)

Identifiants chimiques

64-18-6
46.025
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
CHEBI:30751
C(=O)O
CH2O2
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
formic acid
missing translation for 'recommendations'

missing translation for 'recommendations'

Videos
Sécurité et manipulation

Sécurité et manipulation

Danger

FDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only