Learn More
![](http://beta-static.fishersci.ca/images/Acid.gif)
![](http://beta-static.fishersci.ca/images/White.gif)
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Suitable as a mobile phase additive for LC/MS and UHPLC/MS
Supplier: Thermo Fisher Scientific A11710X1AMP
Specifications
0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm | |
2.1 | |
64-18-6 | |
Colorless | |
CH2O2 | |
15, 4269 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.02 | |
OPTIMA™ LC-MS | |
10 x 1 mL | |
Liquid | |
44mbar at 20°C |
Formic Acid | |
8°C | |
101°C | |
≥99.0 % | |
HCOOH | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
≥99.0% | |
1.220g/cm³ | |
Ampules | |
1ppm max. |
Chemical Identifiers
64-18-6 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Acid.gif)
![](http://beta-static.fishersci.ca/images/White.gif)
DANGER!
Emergency Overview
Flammable liquid and vapor. Causes severe burns by all exposure routes. Hygroscopic. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:3
Flammability:2
Instability:0
DOTInformation : DOT Class 8, : Corrosive