Learn More
![](http://beta-static.fishersci.ca/images/Acid.gif)
![](http://beta-static.fishersci.ca/images/White.gif)
Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™
Suitable as a mobile phase additive for LC/MS and UHPLC/MS
$245.00 - $1329.50
Chemical Identifiers
CAS | 64-18-6 |
---|---|
Molecular Formula | CH2O2 |
InChI Key | BDAGIHXWWSANSR-UHFFFAOYSA-N |
Synonym | methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt |
PubChem CID | 284 |
ChEBI | CHEBI:30751 |
IUPAC Name | formic acid |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
A11705AMP
|
Thermo Fisher Scientific
A11705AMP |
0.5 mL | Ampule |
Each for $245.00
|
|
||||
A1171AMP
|
Thermo Fisher Scientific
A1171AMP |
1 mL | Ampule |
Each for $274.00
|
|
||||
A1172AMP
|
Thermo Fisher Scientific
A1172AMP |
2 mL | Ampule |
Each for $282.50
|
|
||||
A11710X1AMP
|
Thermo Fisher Scientific
A11710X1AMP |
10 x 1 mL | Ampules |
Each for $1,329.50
|
|
||||
A11750
![]() |
Thermo Fisher Scientific
A11750 |
50 mL | Nalgene™ Poly Bottle |
Each for $318.33
|
![]() |
||||
Chemical Identifiers
64-18-6 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
284 | |
formic acid |
CH2O2 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
CHEBI:30751 |
Specifications
0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm | |
8°C | |
101°C | |
≥99.0 % | |
HCOOH | |
15, 4269 | |
BDAGIHXWWSANSR-UHFFFAOYSA-N | |
formic acid | |
284 | |
46.02 | |
OPTIMA™ LC-MS | |
0.5 mL | |
Liquid | |
44mbar at 20°C |
2.1 | |
64-18-6 | |
Colorless | |
CH2O2 | |
3297 | |
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt | |
C(=O)O | |
46.025 | |
CHEBI:30751 | |
≥99.0% | |
1.220g/cm³ | |
Ampule | |
1ppm max. | |
Formic Acid |
Safety and Handling
![](http://beta-static.fishersci.ca/images/Acid.gif)
![](http://beta-static.fishersci.ca/images/White.gif)
DANGER!
Emergency Overview
Flammable liquid and vapor. Causes severe burns by all exposure routes. Hygroscopic. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.
NFPA
Health:3
Flammability:2
Instability:0
DOTInformation : DOT Class 8, : Corrosive