missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Formic Acid, 99.0+%, Optima™ LC/MS Grade, Fisher Chemical™ Edge
Edge

Suitable as a mobile phase additive for LC/MS and UHPLC/MS

Supplier:  Thermo Fisher Scientific A11750

 View more versions of this product

Catalog No. A11750


Edge
Explore available promotions

Description

Description

Specifications

Specifications

0.01 AU max. at 254nm, 0.55 AU max. at 230nm, 0.85 AU max. at 220nm, 1.25 AU max. at 210nm
2.1
64-18-6
Colorless
CH2O2
3297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
C(=O)O
46.025
CHEBI:30751
≥99.0%
1.220g/cm³
Nalgene™ Poly Bottle
1ppm max.
Formic Acid
8°C
101°C
≥99.0 %
HCOOH
15, 4269
BDAGIHXWWSANSR-UHFFFAOYSA-N
formic acid
284
46.02
OPTIMA™ LC-MS
50 mL
Liquid
44mbar at 20°C

Chemical Identifiers

64-18-6
3297
methanoic acid, formylic acid, aminic acid, bilorin, hydrogen carboxylic acid, formisoton, myrmicyl, formira, acide formique, collo-bueglatt
CHEBI:30751
CH2O2
BDAGIHXWWSANSR-UHFFFAOYSA-N
284
formic acid
Videos
Safety and Handling

Safety and Handling

DANGER!

Emergency Overview
Flammable liquid and vapor. Causes severe burns by all exposure routes. Hygroscopic. Wear personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required. Rinse immediately with plenty of water, also under the eyelids, for at least 15 minutes. Immediate medical attention is required. Move to fresh air. If breathing is difficult, give oxygen. If breathing is difficult, give oxygen. Do not use mouth-to-mouth resuscitation if victim ingested or inhaled the substance; induce artifici.

NFPA

Health:3
Flammability:2
Instability:0

DOTInformation : DOT Class 8, : Corrosive

SDS Documents

Documents

Promotions

Promotions