missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bulwark™ FR Lab Coats, Knit Cuffs
Lab coat designed for functionality, comfort, and safety with a CAT 1 arc rating and NFPA 2112 compliance.
Supplier: Bulwark™ KNC2RBLG0L
Description
- Certifications Level 1: NFPA 2112, ASTM F1506
- Certifications Level 2: Category 1 Protection
- Hazard: Arc Flash, Flash Fire
- Wash Care: Industrial Laundry - Light Soil
- Rib cuffs for protection during movement
- Concealed snap front closure to avoid scratching surfaces
- Four front mitered style pockets for ample storage
- Pass-through at hips to access underneath
- Protection: ATPV Arc Rating: 4.4
- Country of Origin: Imported, Made in USA Fabric
Specifications
Flame Resistant Lab Coat | |
Men's | |
Royal Blue | |
Knit | |
Non-sterile | |
Tall | |
KNC2RB | |
NFPA 2112, ASTM 1506 |
Large | |
Nomex | |
Snaps | |
Reusable | |
Flame | |
4 | |
4.4 Calories/cm2 | |
Sewn |