missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Bulwark™ FR Lab Coats, Knit Cuffs
Lab coat designed for functionality, comfort, and safety with a CAT 1 arc rating and NFPA 2112 compliance.
$228.72 - $308.77
Specifications
Product Type | Flame Resistant Lab Coat |
---|---|
Gender | Men's |
Material | Nomex |
Color | Royal Blue |
Closure Type | Snaps |
Catalog Number | Mfr. No. | Size | Style | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Style | Price | Quantity | ||||
17004364
|
Bulwark™
KNC2RBXS0R |
X-Small | Regular |
Each for $228.72
|
|
||||
17004365
|
Bulwark™
KNC2RBSM0R |
Small | Regular |
Each for $228.72
|
|
||||
17004366
|
Bulwark™
KNC2RBMD0R |
Medium | Regular |
Each for $228.72
|
|
||||
17004367
|
Bulwark™
KNC2RBLG0R |
Large | Regular |
Each for $228.72
|
|
||||
17004368
|
Bulwark™
KNC2RBXL0R |
X-Large | Regular |
Each for $228.72
|
|
||||
17004369
|
Bulwark™
KNC2RB2L0R |
2X-Large | Regular |
Each for $228.72
|
|
||||
17004370
|
Bulwark™
KNC2RB3L0R |
3X-Large | Regular |
Each for $228.72
|
|
||||
17004371
|
Bulwark™
KNC2RB4L0R |
4X-Large | Regular |
Each for $228.72
|
|
||||
17004372
|
Bulwark™
KNC2RB5L0R |
5X-Large | Regular |
Each for $228.72
|
|
||||
17004373
|
Bulwark™
KNC2RBMD0L |
Medium | Tall |
Each for $308.77
|
|
||||
17004374
|
Bulwark™
KNC2RBLG0L |
Large | Tall |
Each for $308.77
|
|
||||
17004375
|
Bulwark™
KNC2RBXL0L |
X-Large | Tall |
Each for $308.77
|
|
||||
17004376
|
Bulwark™
KNC2RB2L0L |
2X-Large | Tall |
Each for $308.77
|
|
||||
17004377
|
Bulwark™
KNC2RB3L0L |
3X-Large | Tall |
Each for $308.77
|
|
||||
17004378
|
Bulwark™
KNC2RB4L0L |
4X-Large | Tall |
Each for $308.77
|
|
||||
17004379
|
Bulwark™
KNC2RB5L0L |
5X-Large | Tall |
Each for $308.77
|
|
||||
Description
- Certifications Level 1: NFPA 2112, ASTM F1506
- Certifications Level 2: Category 1 Protection
- Hazard: Arc Flash, Flash Fire
- Wash Care: Industrial Laundry - Light Soil
- Rib cuffs for protection during movement
- Concealed snap front closure to avoid scratching surfaces
- Four front mitered style pockets for ample storage
- Pass-through at hips to access underneath
- Protection: ATPV Arc Rating: 4.4
- Country of Origin: Imported, Made in USA Fabric
Specifications
Flame Resistant Lab Coat | |
Nomex | |
Snaps | |
Reusable | |
Flame | |
KNC2RB | |
NFPA 2112, ASTM 1506 |
Men's | |
Royal Blue | |
Knit | |
Non-sterile | |
4 | |
4.4 Calories/cm2 | |
Sewn |