Thermo Scientific™ Freezer and Refrigerator Shelves
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
Supplier:  Thermo Scientific™ 7125
 View more versions of this product
Catalog No. 12-370-653
Explore available promotions
Description
Description
Maximize lab storage with Thermo Scientific™ Freezer and Refrigerator Shelves, designed for use as replacement shelves in high-performance laboratory refrigerators, freezers and ULTs.
  • Epoxy-coated open-wire shelves
  • Full- and half-depth sizes
  • Undercounter units use full-depth shelves only
Specifications
Specifications
Epoxy-coated open wire shelves
11.5 cu. ft.
Epoxy-coated Steel
4.7 cu. ft. (133L), 11.5 cu. ft. (326L) units
General Freezer Items
Epoxy-Coated Open Wire Shelves
326 L
Plastic
Warranty varies by country, please contact us for details