missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kappler™ Frontline™ 500 Vapor Total Encapsulating Suit
Broad chemical holdout plus flame resistance and radiant heat protection for additional hazards encountered in a chemical flash-fire
Supplier: Kappler™ F5H582SILGXL91
Description
- Seams are sewn and double heat-sealed/taped for added protection
- Large expanded-view face shield improves field of vision
- 122cm (48 in.) Gas-tight PVC zipper
- Double storm flap with hook and loop closure
- Attached sock boots with boot flap
- Field replaceable 2N1™ glove system includes butyl gloves with specially attached barrier glove liner and knitted Kevlar™ overglove
- Silver with high-vis orange taped seam
- Front entry
- Two exhalation valves
- Hazmat response
- Chemical handling with potential for chemical flash-fire
Certifications
Certified to NFPA 1991
Specifications
NFPA 1991 | |
Petrochemical operations, line maintenance, tank cleaning, refueling situations, hazmat response, chemical handling with potential for chemical flash-fire | |
Large/X-Large | |
Expanded | |
Attached gloves | |
Sealed, Taped |
Silver | |
Frontline™ 500 FR | |
Attached boots, Boot overflaps | |
Hook and loop | |
Front entry, expanded back, double-taped seams, 2 exhalation valves, double storm flap with hook and loop closure, attached sock boots with boot flap,butyl gloves with specially attached barrier glove liner and knitted Dupont™ Kevlar™ aramid fiber overglove |