missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Kappler™ Frontline™ 500 Vapor Total Encapsulating Suit
Broad chemical holdout plus flame resistance and radiant heat protection for additional hazards encountered in a chemical flash-fire
$7919.26 - $7919.26
Specifications
Certifications/Compliance | NFPA 1991 |
---|---|
Color | Silver |
For Use With (Application) | Petrochemical operations, line maintenance, tank cleaning, refueling situations, hazmat response, chemical handling with potential for chemical flash-fire |
Material | Frontline™ 500 FR |
Ankle Style | Attached boots, Boot overflaps |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
18-025-691
|
Kappler™
F5H582SILGXL91 |
Large/X-Large |
N/A
|
N/A | |||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
- Seams are sewn and double heat-sealed/taped for added protection
- Large expanded-view face shield improves field of vision
- 122cm (48 in.) Gas-tight PVC zipper
- Double storm flap with hook and loop closure
- Attached sock boots with boot flap
- Field replaceable 2N1™ glove system includes butyl gloves with specially attached barrier glove liner and knitted Kevlar™ overglove
- Silver with high-vis orange taped seam
- Front entry
- Two exhalation valves
- Hazmat response
- Chemical handling with potential for chemical flash-fire
Certifications
Certified to NFPA 1991
Specifications
NFPA 1991 | |
Petrochemical operations, line maintenance, tank cleaning, refueling situations, hazmat response, chemical handling with potential for chemical flash-fire | |
Attached boots, Boot overflaps | |
Hook and loop | |
Front entry, expanded back, double-taped seams, 2 exhalation valves, double storm flap with hook and loop closure, attached sock boots with boot flap,butyl gloves with specially attached barrier glove liner and knitted Dupont™ Kevlar™ aramid fiber overglove |
Silver | |
Frontline™ 500 FR | |
Expanded | |
Attached gloves | |
Sealed, Taped |