Learn More
Fisherbrand™ Full- and Mid-Size Faceshields Edge
Edge
High-quality, optically clear, distortion-free 7mil film
Supplier:  Fisherbrand™ 4201F
 Afficher plus de versions de ce produit
Numéro de catalogue. 19-460-102
Il en reste null
Edge
Découvrez plus d'offres spéciales
Description
Description
  • Polyester lens is treated with antifog and antistatic coating
  • Headpiece is held in place by a stretch strap and includes a latex-free foam band for comfortable extended wear
Compliance
Conform to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and the CDC's recommendations for protective eyewear.
Spécifications
Spécifications
Anti-fog, Anti-static
Clear
Polyester
Conforms to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and the CDC's recommendation for protective eyewear.
Full-Faceshield
missing translation for 'recommendations'
missing translation for 'recommendations'
Videos
FDS