Fisherbrand™ Full- and Mid-Size Faceshields
High-quality, optically clear, distortion-free 7mil film
$46.61 - $46.61
Specifications
Certifications/Compliance | Conforms to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and the CDC's recommendation for protective eyewear. |
---|---|
Lens Tint | Clear |
Lens Material | Polyester |
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
19-460-102
![]() |
Fisherbrand™
4201F |
Full-Faceshield |
Pack of 25 Each for $46.61
Case of 4 Pack for $186.44
|
![]() |
|||||
Description
- Polyester lens is treated with antifog and antistatic coating
- Headpiece is held in place by a stretch strap and includes a latex-free foam band for comfortable extended wear
Compliance
Conform to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and the CDC's recommendation for protective eyewear.
Specifications
Conforms to OSHA 29CFR Part 1910.1030 for bloodborne pathogens and the CDC's recommendation for protective eyewear. | |
Clear | |
Polyester |