missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furfuryl isocyanate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 456490050

 View more versions of this product

Catalog No. AC456490050


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Furfuryl isocyanate
157.0°C to 158.0°C
54.5°C
96% min. (GC)
MFCD03180268
1.148
UIADMYLYGJYUSQ-UHFFFAOYSA-N
2-(isocyanatomethyl)furan
4223811
97%
71189-15-6
1.1480g/mL
Authentic
C6H5NO2
5 g
2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl
O=C=NCC1=CC=CO1
123.11
123.11

Chemical Identifiers

71189-15-6
123.11
UIADMYLYGJYUSQ-UHFFFAOYSA-N
4223811
O=C=NCC1=CC=CO1
C6H5NO2
MFCD03180268
2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl
2-(isocyanatomethyl)furan
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause allergy or asthma symptoms or

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and k

GHS Signal Word: Danger

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only