Learn More
Furfuryl isocyanate, 97%
CAS: 71189-15-6 | C6H5NO2 | 123.11 g/mol
$351.72 - $351.72
Chemical Identifiers
| CAS | 71189-15-6 |
|---|---|
| Molecular Formula | C6H5NO2 |
| Molecular Weight (g/mol) | 123.11 |
| MDL Number | MFCD03180268 |
| InChI Key | UIADMYLYGJYUSQ-UHFFFAOYSA-N |
| Synonym | 2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl |
| PubChem CID | 4223811 |
| IUPAC Name | 2-(isocyanatomethyl)furan |
| SMILES | O=C=NCC1=CC=CO1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AC456490050
|
Thermo Scientific Chemicals
456490050 |
5 g |
Each for $351.72
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 71189-15-6 | |
| 123.11 | |
| UIADMYLYGJYUSQ-UHFFFAOYSA-N | |
| 4223811 | |
| O=C=NCC1=CC=CO1 |
| C6H5NO2 | |
| MFCD03180268 | |
| 2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl | |
| 2-(isocyanatomethyl)furan |
Specifications
| 71189-15-6 | |
| 1.1480g/mL | |
| Authentic | |
| C6H5NO2 | |
| 5 g | |
| 2-isocyanatomethyl furan, furfuryl isocyanate, 2-furylmethyl isocyanate, furfurylisocyanate, furylmethylisocyanate, 2-isocyanatomethyl-furan, 2-furyl methyl isocyanate, furan, 2-isocyanatomethyl | |
| O=C=NCC1=CC=CO1 | |
| 123.11 | |
| 123.11 | |
| Furfuryl isocyanate |
| 157.0°C to 158.0°C | |
| 54.5°C | |
| 96% min. (GC) | |
| MFCD03180268 | |
| 1.148 | |
| UIADMYLYGJYUSQ-UHFFFAOYSA-N | |
| 2-(isocyanatomethyl)furan | |
| 4223811 | |
| 97% |
Safety and Handling
GHS H Statement
Flammable liquid and vapour.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause allergy or asthma symptoms or
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN: Wash with plenty of soap and water.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove to fresh air and k
GHS Signal Word: Danger
RUO – Research Use Only