missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Furo[3,2-b]pyridine, 95%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals H3296603

 View more versions of this product

Catalog No. AAH3296603


Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Furo[3,2-b]pyridine
79°C to 80°C (12 mmHg)
1.5753
1 g
YRTCKZIKGWZNCU-UHFFFAOYSA-N
furo[3,2-b]pyridine
12210217
98%
272-62-8
C7H5NO
MFCD11870744
furo 3,2-b pyridine, 4-azabenzo b furan, furopyridine, furo 3,2-b pyridin, 4-aza-1-benzofuran
O1C=CC2=NC=CC=C12
119.12
119.12

Chemical Identifiers

272-62-8
119.12
YRTCKZIKGWZNCU-UHFFFAOYSA-N
12210217
O1C=CC2=NC=CC=C12
C7H5NO
MFCD11870744
furo 3,2-b pyridine, 4-azabenzo b furan, furopyridine, furo 3,2-b pyridin, 4-aza-1-benzofuran
furo[3,2-b]pyridine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only