Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
missing translation for 'pleaseLogIn'
Furo[3,2-b]pyridine, 95%, Thermo Scientific Chemicals
$442.68 - $1464.74
Chemical Identifiers
CAS | 272-62-8 |
---|---|
Molecular Formula | C7H5NO |
Molecular Weight (g/mol) | 119.12 |
MDL Number | MFCD11870744 |
InChI Key | YRTCKZIKGWZNCU-UHFFFAOYSA-N |
Synonym | furo 3,2-b pyridine, 4-azabenzo b furan, furopyridine, furo 3,2-b pyridin, 4-aza-1-benzofuran |
PubChem CID | 12210217 |
IUPAC Name | furo[3,2-b]pyridine |
SMILES | O1C=CC2=NC=CC=C12 |
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Qty | |||||
AAH3296603
|
Thermo Scientific Chemicals
H3296603 |
1 g |
chaque for $442.68
|
|
|||||
AAH3296606
|
Thermo Scientific Chemicals
H3296606 |
5 g |
chaque for $1,464.74
|
|
|||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
272-62-8 | |
119.12 | |
YRTCKZIKGWZNCU-UHFFFAOYSA-N | |
12210217 | |
O1C=CC2=NC=CC=C12 |
C7H5NO | |
MFCD11870744 | |
furo 3,2-b pyridine, 4-azabenzo b furan, furopyridine, furo 3,2-b pyridin, 4-aza-1-benzofuran | |
furo[3,2-b]pyridine |
Specifications
272-62-8 | |
C7H5NO | |
MFCD11870744 | |
furo 3,2-b pyridine, 4-azabenzo b furan, furopyridine, furo 3,2-b pyridin, 4-aza-1-benzofuran | |
O1C=CC2=NC=CC=C12 | |
119.12 | |
119.12 | |
Furo[3,2-b]pyridine |
79°C to 80°C (12 mmHg) | |
1.5753 | |
1 g | |
YRTCKZIKGWZNCU-UHFFFAOYSA-N | |
furo[3,2-b]pyridine | |
12210217 | |
98% |
Safety and Handling
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only