missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gadolinium silicide, 99.9% (metals basis), Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 3635604

 View more versions of this product

Catalog No. AA3635604


Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Gadolinium silicide
Pieces
(metals basis)
MFCD00144382
[SiH4].[SiH4].[Gd+3]
221.484
213.42
Odorless
12134-75-7
2 g
GdH8Si2+3
AOLGUBMBPNHJFD-UHFFFAOYSA-N
gadolinium(3+);silane
71434213
99.9%

Chemical Identifiers

12134-75-7
221.484
AOLGUBMBPNHJFD-UHFFFAOYSA-N
gadolinium(3+);silane
GdH8Si2+3
MFCD00144382
71434213
[SiH4].[SiH4].[Gd+3]
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 235-216-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions