missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
gamma-Linolenic Acid 98.0+%, TCI America™
Supplier: TCI America L01521G
Specifications
gamma-Linolenic Acid | |
Yellow | |
MFCD00065718 | |
gamma-linolenic acid, gamolenic acid, 6z,9z,12z-octadeca-6,9,12-trienoic acid, z,z,z-6,9,12-octadecatrienoic acid, ccris 7668, ligla, unii-78yc2max4o, acide gamolenique french, acido gamolenico spanish, acidum gamolenicum latin | |
CCCCC\C=C/C\C=C/C\C=C/CCCCC(O)=O | |
278.44 | |
CHEBI:28661 | |
≥98.0% (GC,T) |
506-26-3 | |
C18H30O2 | |
1 g | |
VZCCETWTMQHEPK-QNEBEIHSSA-N | |
(6Z,9Z,12Z)-octadeca-6,9,12-trienoic acid | |
5280933 | |
278.44 | |
Liquid |
Chemical Identifiers
506-26-3 | |
278.44 | |
VZCCETWTMQHEPK-QNEBEIHSSA-N | |
5280933 | |
(6Z,9Z,12Z)-octadeca-6,9,12-trienoic acid |
C18H30O2 | |
MFCD00065718 | |
gamma-linolenic acid, gamolenic acid, 6z,9z,12z-octadeca-6,9,12-trienoic acid, z,z,z-6,9,12-octadecatrienoic acid, ccris 7668, ligla, unii-78yc2max4o, acide gamolenique french, acido gamolenico spanish, acidum gamolenicum latin | |
CHEBI:28661 | |
CCCCC\C=C/C\C=C/C\C=C/CCCCC(O)=O |
Safety and Handling
EINECSNumber : (2)-0609
TSCA : No
Recommended Storage : Refrigerator