missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
gamma-Linolenic Acid 98.0+%, TCI America™
$169.56 - $839.10
Chemical Identifiers
CAS | 506-26-3 |
---|---|
Molecular Formula | C18H30O2 |
Molecular Weight (g/mol) | 278.44 |
MDL Number | MFCD00065718 |
InChI Key | VZCCETWTMQHEPK-QNEBEIHSSA-N |
Synonym | gamma-linolenic acid, gamolenic acid, 6z,9z,12z-octadeca-6,9,12-trienoic acid, z,z,z-6,9,12-octadecatrienoic acid, ccris 7668, ligla, unii-78yc2max4o, acide gamolenique french, acido gamolenico spanish, acidum gamolenicum latin |
PubChem CID | 5280933 |
ChEBI | CHEBI:28661 |
IUPAC Name | (6Z,9Z,12Z)-octadeca-6,9,12-trienoic acid |
SMILES | CCCCC\C=C/C\C=C/C\C=C/CCCCC(O)=O |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
L0152100MG
|
TCI America
L0152100MG |
100 mg |
Each for $169.56
|
|
|||||
L01521G
|
TCI America
L01521G |
1 g |
Each for $839.10
|
|
|||||
Chemical Identifiers
506-26-3 | |
278.44 | |
VZCCETWTMQHEPK-QNEBEIHSSA-N | |
5280933 | |
(6Z,9Z,12Z)-octadeca-6,9,12-trienoic acid |
C18H30O2 | |
MFCD00065718 | |
gamma-linolenic acid, gamolenic acid, 6z,9z,12z-octadeca-6,9,12-trienoic acid, z,z,z-6,9,12-octadecatrienoic acid, ccris 7668, ligla, unii-78yc2max4o, acide gamolenique french, acido gamolenico spanish, acidum gamolenicum latin | |
CHEBI:28661 | |
CCCCC\C=C/C\C=C/C\C=C/CCCCC(O)=O |
Specifications
506-26-3 | |
C18H30O2 | |
100 mg | |
VZCCETWTMQHEPK-QNEBEIHSSA-N | |
(6Z,9Z,12Z)-octadeca-6,9,12-trienoic acid | |
5280933 | |
278.44 | |
Liquid |
Yellow | |
MFCD00065718 | |
gamma-linolenic acid, gamolenic acid, 6z,9z,12z-octadeca-6,9,12-trienoic acid, z,z,z-6,9,12-octadecatrienoic acid, ccris 7668, ligla, unii-78yc2max4o, acide gamolenique french, acido gamolenico spanish, acidum gamolenicum latin | |
CCCCC\C=C/C\C=C/C\C=C/CCCCC(O)=O | |
278.44 | |
CHEBI:28661 | |
≥98.0% (GC,T) | |
gamma-Linolenic Acid |
Safety and Handling
EINECSNumber : (2)-0609
TSCA : No
Recommended Storage : Refrigerator