Covidien General Purpose Specimen Containers
4 oz. (120mL) specimen collection containers
 View more versions of this product
Catalog No. 14-375-149
Explore available promotions
Description
Description
  • Full-turn screw thread provides excellent leak-resistant seal
  • Translucent polypropylene is tough, durable, nonbreakable, and inert; tolerates strong acids, bases, and most organic solvents
  • Sterile and nonsterile varieties available - sterile containers have integrity seal or O.R. package
  • Cups are graduated in both U.S. and metric scale
Compliance
NCCLS Specimen Handling Guidelines for primary containers (M29-T2, ol.11, No.14)
Specifications
Specifications
Yes
Sterile
120 mL
Green
Translucent
Polypropylene
4 oz.
NCCLS Specimen Handling Guidelines for primary containers (M29-T2, ol.11, No.14)
Integrity Seal
Bulk, 100/Cs.