missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Covidien General Purpose Specimen Containers
4 oz. (120mL) specimen collection containers
Description
- Full-turn screw thread provides excellent leak-resistant seal
- Translucent polypropylene is tough, durable, nonbreakable, and inert; tolerates strong acids, bases, and most organic solvents
- Sterile and nonsterile varieties available - sterile containers have integrity seal or O.R. package
- Cups are graduated in both U.S. and metric scale
Compliance
NCCLS Specimen Handling Guidelines for primary containers (M29-T2, ol.11, No.14)
Specifications
Yes | |
Sterile | |
120 mL | |
Green | |
Translucent |
Polypropylene | |
4 oz. | |
NCCLS Specimen Handling Guidelines for primary containers (M29-T2, ol.11, No.14) | |
Integrity Seal | |
Individually wrapped, 100/Cs. |