missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Genistein, 99%, synthetic, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 328271000
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Genistein | |
446-72-0 | |
White to Beige | |
98.5% min. (HPLC) | |
C15H10O5 | |
100 mg | |
genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein | |
TZBJGXHYKVUXJN-UHFFFAOYSA-N | |
5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one | |
5280961 | |
270.2 | |
Crystalline Powder |
Synthetic | |
297.0°C to 298.0°C | |
Authentic | |
Glass bottle | |
MFCD00016952 | |
15, 4426 | |
Solubility in water: slightly soluble. Other solubilities: soluble in dmso and ethanol,soluble in usual organic solvents,soluble in diluted alkalies with yellow color | |
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O | |
270.24 | |
CHEBI:28088 | |
99% |
Chemical Identifiers
446-72-0 | |
270.24 | |
TZBJGXHYKVUXJN-UHFFFAOYSA-N | |
5280961 | |
5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one |
C15H10O5 | |
MFCD00016952 | |
genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein | |
CHEBI:28088 | |
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O |
Safety and Handling
GHS H Statement
Harmful if swallowed.
GHS Signal Word: Warning
EINECSNumber : 207-174-9
RUO – Research Use Only