missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Genistein, 99%, synthetic, Thermo Scientific Chemicals

$211.24 - $598.53

Chemical Identifiers

CAS 446-72-0
Molecular Formula C15H10O5
Molecular Weight (g/mol) 270.24
MDL Number MFCD00016952
InChI Key TZBJGXHYKVUXJN-UHFFFAOYSA-N
Synonym genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein
PubChem CID 5280961
ChEBI CHEBI:28088
IUPAC Name 5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one
SMILES OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC328270250
SDS View Documents
Thermo Scientific Chemicals
328270250
25 mg Glass bottle
Each for $211.24
 
AC328271000
SDS View Documents
Thermo Scientific Chemicals
328271000
100 mg Glass bottle
Each for $598.53
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

446-72-0
270.24
TZBJGXHYKVUXJN-UHFFFAOYSA-N
5280961
5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one
C15H10O5
MFCD00016952
genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein
CHEBI:28088
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O

Specifications

446-72-0
White to Beige
98.5% min. (HPLC)
C15H10O5
25 mg
genistein, prunetol, genisteol, 4',5,7-trihydroxyisoflavone, genisterin, sophoricol, 5,7,4'-trihydroxyisoflavone, bonistein, 5,7-dihydroxy-3-4-hydroxyphenyl-4h-chromen-4-one, genestein
TZBJGXHYKVUXJN-UHFFFAOYSA-N
5,7-dihydroxy-3-(4-hydroxyphenyl)chromen-4-one
5280961
270.2
Crystalline Powder
297.0°C to 298.0°C
Authentic
Glass bottle
MFCD00016952
15, 4426
Solubility in water: slightly soluble. Other solubilities: soluble in dmso and ethanol,soluble in usual organic solvents,soluble in diluted alkalies with yellow color
OC1=CC=C(C=C1)C1=COC2=CC(O)=CC(O)=C2C1=O
270.24
CHEBI:28088
99%
Genistein, Synthetic
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.

GHS Signal Word: Warning

EINECSNumber : 207-174-9

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only