Amsbio LLC (formerly Genlantis) GenePORTER™ 3000 Reagent
Provides high transfection efficiency for hard to transfect cells
$1665.92 - $10926.83
Specifications
For Use With (Application) | Proprietary cationic Lipid that utilizes advanced conjugation enhancement (ACE) technology to maximize target molecule delivery with minimal cytotoxicity |
---|
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Quantity | Price | Quantity | ||||
T203115
|
Amsbio LLC (formerly Genlantis)
T203115 |
15mL GenePORTER™ 3000 reagent, 1200mL GP3K diluents | 15 mL |
N/A
|
N/A | ||||
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
T203015
|
Amsbio LLC (formerly Genlantis)
T203015 |
1.5mL GenePORTER™ 3000 reagent, 2 x 6mL GP3K diluents | 1.5 mL |
Each for $1,665.92
|
|
||||
Description
The GenePORTER 3000™ Transfection Reagent is a proprietary cationic lipid formulation that utilizes Advanced Carrier Enhancement (ACE) technology to maximize target molecule delivery with minimal cytotoxicity.
- Designed specifically for difficult-to-transfect cells
- Rapid protocol: Optimized for same day transfection
- Superior performance in both serum-free and serum-containing media
Specifications
Proprietary cationic Lipid that utilizes advanced conjugation enhancement (ACE) technology to maximize target molecule delivery with minimal cytotoxicity |