Amsbio LLC (formerly Genlantis) GenePORTER™ 3000 Reagent DFS Item
Provides high transfection efficiency for hard to transfect cells
Supplier:  Amsbio LLC (formerly Genlantis) T203115
 View more versions of this product
Catalog No. T203115
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Explore available promotions
Description
Description
The GenePORTER 3000™ Transfection Reagent is a proprietary cationic lipid formulation that utilizes Advanced Carrier Enhancement (ACE) technology to maximize target molecule delivery with minimal cytotoxicity.
  • Designed specifically for difficult-to-transfect cells
  • Rapid protocol: Optimized for same day transfection
  • Superior performance in both serum-free and serum-containing media
Specifications
Specifications
Proprietary cationic Lipid that utilizes advanced conjugation enhancement (ACE) technology to maximize target molecule delivery with minimal cytotoxicity
15mL GenePORTER™ 3000 reagent, 1200mL GP3K diluents
15 mL