missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Geranyl bromide, 96%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals A1409322

 View more versions of this product

Catalog No. AAA1409322


Explore available promotions

Description

Description

Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Specifications

Geranyl bromide
1.094
95°C (203°F)
1.504
100 g
Light sensitive
Immiscible with water.
CC(=CCCC(=CCBr)C)C
217.15
217.16
6138-90-5
99°C to 101°C (10 mmHg)
C10H17Br
MFCD00000243
1422412
geranyl bromide, e-1-bromo-3,7-dimethylocta-2,6-diene, 2,6-octadiene, 1-bromo-3,7-dimethyl-, e, 2,6-octadiene, 1-bromo-3,7-dimethyl-, 2e, geranylbromide, 2e-1-bromo-3,7-dimethyl-2,6-octadiene, 6e-8-bromo-2,6-dimethylocta-2,6-diene, 1-bromo-3,7-dimethyl-2,6-octadiene, trans-geranyl bromide, e-geranyl bromide
JSCUZAYKVZXKQE-JXMROGBWSA-N
(2E)-1-bromo-3,7-dimethylocta-2,6-diene
5365867
96%

Chemical Identifiers

6138-90-5
217.15
JSCUZAYKVZXKQE-JXMROGBWSA-N
5365867
CC(=CCCC(=CCBr)C)C
C10H17Br
MFCD00000243
geranyl bromide, e-1-bromo-3,7-dimethylocta-2,6-diene, 2,6-octadiene, 1-bromo-3,7-dimethyl-, e, 2,6-octadiene, 1-bromo-3,7-dimethyl-, 2e, geranylbromide, 2e-1-bromo-3,7-dimethyl-2,6-octadiene, 6e-8-bromo-2,6-dimethylocta-2,6-diene, 1-bromo-3,7-dimethyl-2,6-octadiene, trans-geranyl bromide, e-geranyl bromide
(2E)-1-bromo-3,7-dimethylocta-2,6-diene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H318-H335

EINECSNumber : 228-123-7

TSCA : Yes

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only