missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Geranyl bromide, 96%, Thermo Scientific Chemicals

$108.29 - $1092.05

Chemical Identifiers

CAS 6138-90-5
Molecular Formula C10H17Br
Molecular Weight (g/mol) 217.15
MDL Number MFCD00000243
InChI Key JSCUZAYKVZXKQE-JXMROGBWSA-N
Synonym geranyl bromide, e-1-bromo-3,7-dimethylocta-2,6-diene, 2,6-octadiene, 1-bromo-3,7-dimethyl-, e, 2,6-octadiene, 1-bromo-3,7-dimethyl-, 2e, geranylbromide, 2e-1-bromo-3,7-dimethyl-2,6-octadiene, 6e-8-bromo-2,6-dimethylocta-2,6-diene, 1-bromo-3,7-dimethyl-2,6-octadiene, trans-geranyl bromide, e-geranyl bromide
PubChem CID 5365867
IUPAC Name (2E)-1-bromo-3,7-dimethylocta-2,6-diene
SMILES CC(=CCCC(=CCBr)C)C
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAA1409306
View Documents
Thermo Scientific Chemicals
A1409306
5 g
Each for $108.29
 
AAA1409314
View Documents
Thermo Scientific Chemicals
A1409314
25 g
Each for $359.75
 
AAA1409322
View Documents
Thermo Scientific Chemicals
A1409322
100 g
Each for $1,092.05
 
Description

Description

Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Geranyl bromide is used for the preparation of 3,7-dihydroxyflavone derivatives and baicalein. It is also involved in the synthesis of potential flavonoidic modulators of P-glycoprotein activity. Further, it reacts with benzenesulfinic acid to prepare geranyl phenyl sulfone. In addition to this, it takes part in the palladium catalyzed cross coupling with aryl and alkenylgold(I) phosphanes.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with strong oxidizing agents.
Specifications

Chemical Identifiers

6138-90-5
217.15
JSCUZAYKVZXKQE-JXMROGBWSA-N
5365867
CC(=CCCC(=CCBr)C)C
C10H17Br
MFCD00000243
geranyl bromide, e-1-bromo-3,7-dimethylocta-2,6-diene, 2,6-octadiene, 1-bromo-3,7-dimethyl-, e, 2,6-octadiene, 1-bromo-3,7-dimethyl-, 2e, geranylbromide, 2e-1-bromo-3,7-dimethyl-2,6-octadiene, 6e-8-bromo-2,6-dimethylocta-2,6-diene, 1-bromo-3,7-dimethyl-2,6-octadiene, trans-geranyl bromide, e-geranyl bromide
(2E)-1-bromo-3,7-dimethylocta-2,6-diene

Specifications

6138-90-5
99°C to 101°C (10 mmHg)
C10H17Br
MFCD00000243
1422412
geranyl bromide, e-1-bromo-3,7-dimethylocta-2,6-diene, 2,6-octadiene, 1-bromo-3,7-dimethyl-, e, 2,6-octadiene, 1-bromo-3,7-dimethyl-, 2e, geranylbromide, 2e-1-bromo-3,7-dimethyl-2,6-octadiene, 6e-8-bromo-2,6-dimethylocta-2,6-diene, 1-bromo-3,7-dimethyl-2,6-octadiene, trans-geranyl bromide, e-geranyl bromide
JSCUZAYKVZXKQE-JXMROGBWSA-N
(2E)-1-bromo-3,7-dimethylocta-2,6-diene
5365867
96%
1.094
95°C (203°F)
1.504
5 g
Light sensitive
Immiscible with water.
CC(=CCCC(=CCBr)C)C
217.15
217.16
Geranyl bromide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H318-H335-H227-H315
Causes serious eye damage.
May cause respiratory irritation.
Combustible liquid.
Causes skin irritation.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H318-H335

EINECSNumber : 228-123-7

TSCA : Yes

Recommended Storage : Keep cold

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only