Learn More
Germanium dioxide, 99.999%, (trace metal basis), electronic grade, Thermo Scientific Chemicals
$224.83 - $3005.98
Chemical Identifiers
CAS | 1310-53-8 |
---|---|
Molecular Formula | GeO2 |
Molecular Weight (g/mol) | 104.59 |
MDL Number | MFCD00011030 |
InChI Key | YBMRDBCBODYGJE-UHFFFAOYSA-N |
Synonym | germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2 |
PubChem CID | 14796 |
IUPAC Name | dioxogermane |
SMILES | O=[Ge]=O |
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
AC190000100
|
Thermo Scientific Chemicals
190000100 |
10 g | Glass bottle |
Each for $224.83
|
|
||||
AC190000500
|
Thermo Scientific Chemicals
190000500 |
50 g | Glass bottle |
Each for $904.41
|
|
||||
AC190002500
|
Thermo Scientific Chemicals
190002500 |
250 g | Glass bottle |
Each for $3,005.98
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For ultra trace analysisChemical Identifiers
1310-53-8 | |
104.59 | |
YBMRDBCBODYGJE-UHFFFAOYSA-N | |
14796 | |
O=[Ge]=O |
GeO2 | |
MFCD00011030 | |
germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2 | |
dioxogermane |
Specifications
1115.0°C | |
White | |
10 g | |
GeO2 | |
MFCD00011030 | |
germanium dioxide, germania, germanic acid, germanium oxide, germanium iv oxide, germanic oxide crystalline, dioxogermanium, germanic oxide, wln: ge o2 | |
YBMRDBCBODYGJE-UHFFFAOYSA-N | |
dioxogermane | |
14796 | |
99.999% | |
Glass bottle | |
Germanium dioxide |
1310-53-8 | |
Powder | |
99.999% | |
GeO2 | |
15,4443 | |
Solubility in water: 0.4%. Other solubilities: soluble in about 100 parts boiling water, soluble in acids or solutions of the fixed, alkalies | |
O=[Ge]=O | |
104.59 | |
104.59 | |
Trace Metal Basis | |
6.2300g/mL |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWAL
GHS Signal Word: Warning
EINECSNumber : 215-180-8
RUO – Research Use Only