missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ MEM α, no nucleosides DFS Item

MEM α (Minimum Essential Medium α) is widely used for mammalian cell culture as well as selection for transfected DHFR-negative cells.

Supplier:  Gibco™ 12561049

 View more versions of this product

Catalog No. 12561049


Only null left
Explore available promotions

Description

Description

MEM α (Minimum Essential Medium α) is widely used for mammalian cell culture as well as selection for transfected DHFR-negative cells. MEM α can be used with a variety of suspension and adherent mammalian cells, including keratinocytes, primary rat astrocytes, and human melanoma cells. We offer a variety of MEM α modifications for a range of cell culture applications. Find the right formulation using the media selector tool.

This MEM α is modified as follows:

With: Phenol Red, L-glutamine

Without: Ribonucleosides, Deoxyribonucleosides

Using DMEM α

MEM α is a modification of Minimum Essential Medium (MEM) that contains non-essential amino acids, sodium pyruvate, lipoic acid, vitamin B12, biotin, and ascorbic acid. MEM α is available without nucleosides for use as a selection medium for DG44 and other DHFR-negative cells. This product is made with Earle's salts. MEM α contains no proteins, lipids, or growth factors. Therefore, MEM α requires supplementation, commonly with 10% Fetal Bovine Serum (FBS). MEM α uses a sodium bicarbonate buffer system (2.2 g/L), and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Specifications

Specifications

HeLa, BHK-21, 293, HEP-2, HT-1080, MCF-7, and fibroblasts
Animal Origin-free
Liquid
Standard Serum Supplementation
Sterile-filtered
No HEPES, No Ribonucleosides, No Deoxyribonucleosides
Gibco
Room Temperature
Primary Rat Astrocytes
1 X
MEM α (Minimum Essential Medium α)
Sterile-filtered
Low Glucose, Glutamine, Phenol Red, Sodium Pyruvate
cGMP-compliant under the ISO 13485 standard
1000 mL
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.